Learn More
Aviva Systems Biology Corp NCK1 Antibody - N-terminal region (ARP64517_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64517_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NCK1. It was validated on Western Blot. . Alternate names: MGC12668, NCK, NCKalpha, nck-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-288-280