Learn More
ABclonal Technology NCKAP1 Rabbit pAb
SDP
Catalog No. 5000114948
Encompass_Preferred
Contributes to small GTPase binding activity. Involved in Rac protein signal transduction, positive regulation of Arp2/3 complex-mediated actin nucleation, and positive regulation of lamellipodium assembly. Located in extracellular exosome and focal adhesion. Part of SCAR complex.
Catalog No. 50-001-14948 Supplier ABclonal Technology Supplier No. A1222920UL
Contributes to small GTPase binding activity. Involved in Rac protein signal transduction, positive regulation of Arp2/3 complex-mediated actin nucleation, and positive regulation of lamellipodium assembly. Located in extracellular exosome and focal adhesion. Part of SCAR complex.