Learn More
Aviva Systems Biology Corp NCKAP1L Antibody - C-terminal region (ARP47586_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47586_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NCKAP1L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NCKAP1L. Alternate names: HEM1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59599