Learn More
Cell Signaling Technology NckIPSD Monoclonal Antibody, Human, Mouse, 100 μL
SDP
Catalog No. 502730292
Encompass
This NCKIPSD (E6Z3N) Rabbit Monoclonal Antibody is a recombinant primary antibody designed to detect endogenous levels of total NCKIPSD protein. Also known as SPIN90, NCKIPSD is a cytoskeletal adaptor protein essential for organizing the actin cytoskeleton and regulating actin-based cell motility, microtubule acetylation, and signaling within the central nervous system. This antibody is produced by immunizing animals with a synthetic peptide corresponding to residues surrounding Val716 of human NCKIPSD.
  • Validated applications include western blotting and immunoprecipitation
  • Recognizes isoforms 1 and 3 of NCKIPSD protein
  • Species reactivity: Human, Mouse
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide
Catalog No. 50-273-0292 Supplier Cell Signaling Technology Supplier No. 89661S
This NCKIPSD (E6Z3N) Rabbit Monoclonal Antibody is a recombinant primary antibody designed to detect endogenous levels of total NCKIPSD protein. Also known as SPIN90, NCKIPSD is a cytoskeletal adaptor protein essential for organizing the actin cytoskeleton and regulating actin-based cell motility, microtubule acetylation, and signaling within the central nervous system. This antibody is produced by immunizing animals with a synthetic peptide corresponding to residues surrounding Val716 of human NCKIPSD.
  • Validated applications include western blotting and immunoprecipitation
  • Recognizes isoforms 1 and 3 of NCKIPSD protein
  • Species reactivity: Human, Mouse
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide