Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 4691MSM1XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: SPM614. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: All human Cells. Testis, Ovary, Uterus, Endometrial or Hodgkin s lymphoma. Cellular Staining: Nucleoli. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human NCL protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 76kDa.Chromosome: 2q37.1. Gene Symbol: NCL.
Catalog No. 50-188-5004