Learn More
Aviva Systems Biology Corp NCL Antibody - N-terminal region (ARP40583_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40583_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NCL. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human NCL. Alternate names: C23ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-57533