Learn More
Sino Biological NCL Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 102064T36100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human NCL, and purified by antigen affinity chromatography.
Catalog No. 50-224-6296