Learn More
Leica Biosystems NCL-L-P53-CM5P 0.5ML
Supplier:  Leica Biosystems P53CM5PL
Encompass
Antigen Background The accumulation of p53 protein in response to genotoxic stress in vitro is well established and appears to induce growth arrest and apoptosis by the transcriptional regulation of other genes and possibly by other direct mechanisms. Product Specific Information NCL-L-p53-CM5p is specific for mouse and rat p53 protein.
Catalog No. NC2236147