Learn More
Aviva Systems Biology Corp NCOA1 Antibody - middle region (ARP33991_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33991_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NCOA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NCOA1. Alternate names: F-SRC-1, KAT13A, MGC129719, MGC129720, NCoA-1, RIP160, SRC1, bHLHe42, bHLHe74ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55215