Learn More
Aviva Systems Biology Corp NCOA4 Antibody - C-terminal region (ARP35774_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35774_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NCOA4. It was validated on Western Blot. . Alternate names: ARA70, DKFZp762E1112, ELE1, PTC3, RFGApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rat
Catalog No. 50-284-474