Learn More
Aviva Systems Biology Corp Ncoa4 Antibody - N-terminal region (ARP37744_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37744_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ncoa4. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 1110034E15Rik, 4432406M01Rik, AI227008, ARA70, MGC103382, RfgApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-686