Learn More
New England Biolabs, Inc. NcoI – 1000 units
SDP
Catalog No. 50812120
Encompass
A restriction endonuclease that recognizes the sequence C^CATG_G.
Catalog No. 50-812-120 Supplier New England Biolabs, Inc. Supplier No. R0193S
A restriction endonuclease that recognizes the sequence C^CATG_G.