Learn More
New England Biolabs, Inc. NcoI – 5000 units
SDP
Catalog No. 50812118
Encompass
A restriction endonuclease that recognizes the sequence C^CATG_G.
Catalog No. 50-812-118 Supplier New England Biolabs, Inc. Supplier No. R0193L
A restriction endonuclease that recognizes the sequence C^CATG_G.