Learn More
New England Biolabs, Inc. NcoI – 5000 units
SDP
Supplier:  New England Biolabs, Inc. R0193M
Encompass
A restriction endonuclease that recognizes the sequence C^CATG_G.
Catalog No. 50-812-119