Learn More
Aviva Systems Biology Corp NCOR2 Antibody - N-terminal region (ARP32464_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32464_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NCOR2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NCOR2. Alternate names: SMRT, TRAC, CTG26, SMRTE, TRAC1, N-CoR2, TNRC14, TRAC-1, SMAP270, SMRTE-tauApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat
Catalog No. 50-014-54604