Learn More
Cayman Chemical ncortave Acetate 50mg
SDP
Catalog No. 502912491
Encompass_Preferred
A sterol lipid; inhibits VEGF-induced increases in vascular generation, number, and length in isolated rat retinas at 1 µM; sub-tenon administration at 7.5% w/v prevents and reduces prednisolone-induced intraocular pressure in sheep; subconjunctival administration with 2-deoxyglucose decreases tumor growth in a Tg(TagRb)1Plm mouse model of retinoblastoma at 15% w/v
Catalog No. 50-291-2491 Supplier Cayman Chemical Supplier No. 3644150 MG
A sterol lipid; inhibits VEGF-induced increases in vascular generation, number, and length in isolated rat retinas at 1 µM; sub-tenon administration at 7.5% w/v prevents and reduces prednisolone-induced intraocular pressure in sheep; subconjunctival administration with 2-deoxyglucose decreases tumor growth in a Tg(TagRb)1Plm mouse model of retinoblastoma at 15% w/v