Learn More
Sigma Aldrich Fine Chemicals Biosciences Ncotnc acd mononucleot100MG
Catalog No. 502907828
Encompass
Nicotinic acid mononucleotide (NaMN) is formed from nicotinic acid 5-phosphoribosyl-1-pyrophosphate (PRPP) in the Preiss-Handler pathway in the presence of enzyme phosphoribosyltransferase (NaPRT).
Catalog No. 50-290-7828 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. N7764100MG
Nicotinic acid mononucleotide (NaMN) is formed from nicotinic acid 5-phosphoribosyl-1-pyrophosphate (PRPP) in the Preiss-Handler pathway in the presence of enzyme phosphoribosyltransferase (NaPRT).