Learn More
Medchemexpress LLC NCP2 Anchor | 1380600-13-4 | C38H36N4O8 | 100 MG
SDP
Catalog No. 5000373704
Encompass_Preferred
NCP2 Anchor is used in the synthesis of exon jumping oligomer conjugates. These conjugates complement selected target sites in the human anti-muscular atrophy protein gene, inducing exon 52 jumping. It is suitable for research related to muscular dystrophy.
  • Used in the synthesis of exon jumping oligomer conjugates.
  • Oligomer conjugates complement target sites in the human anti-muscular atrophy protein gene.
  • Induces exon 52 jumping.
  • Applicable for research of muscular dystrophy.
Catalog No. 50-003-73704 Supplier Medchemexpress LLC Supplier No. HY153407100MG
NCP2 Anchor is used in the synthesis of exon jumping oligomer conjugates. These conjugates complement selected target sites in the human anti-muscular atrophy protein gene, inducing exon 52 jumping. It is suitable for research related to muscular dystrophy.
  • Used in the synthesis of exon jumping oligomer conjugates.
  • Oligomer conjugates complement target sites in the human anti-muscular atrophy protein gene.
  • Induces exon 52 jumping.
  • Applicable for research of muscular dystrophy.