Learn More
Enterprise Technology Solutions NCR Paper Padding Adhesive, High-Strength, 1 qt. Bottle
SDP
Catalog No. 5000826620
Encompass_Preferred
Fanapart edge padding adhesive allows Appleton carbonless sheets, in the proper sequence, to be automatically fastened together into form sets. Noncoated surfaces of the CB and CF plies are carefully manufactured to precise standards so carbonless sheets will automatically separate into form sets. Designed to allow you to use the form sets approximately 20 minutes after the High Strength Fanapart Adhesive has been applied. Each bottle contains 1 quart.
Catalog No. 50-008-26620 Supplier Enterprise Technology Solutions Supplier No. NCR2116
Fanapart edge padding adhesive allows Appleton carbonless sheets, in the proper sequence, to be automatically fastened together into form sets. Noncoated surfaces of the CB and CF plies are carefully manufactured to precise standards so carbonless sheets will automatically separate into form sets. Designed to allow you to use the form sets approximately 20 minutes after the High Strength Fanapart Adhesive has been applied. Each bottle contains 1 quart.