Learn More
Aviva Systems Biology Corp NCR1 Antibody - N-terminal region (ARP63659_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63659_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NCR1. It was validated on Western Blot. . Alternate names: CD335, FLJ99094, LY94, NK-p46, NKP46ApplicationWB. Species reactivity: Human
Catalog No. 50-288-046