Learn More
ABclonal Technology NCR3 Rabbit mAb
SDP
Supplier:  ABclonal Technology A24295100UL
Encompass_Preferred
The protein encoded by this gene is a natural cytotoxicity receptor (NCR) that may aid NK cells in the lysis of tumor cells The encoded protein interacts with CD3-zeta (CD247) a T-cell receptor A single nucleotide polymorphism in the 5 untranslated region of this gene has been associated with mild malaria suceptibility Three transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-13225