Learn More
Aviva Systems Biology Corp NCRNA00114 Antibody - N-terminal region (ARP45767_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45767_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NCRNA00114. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NCRNA00114. Alternate names: C21orf24, NCRNA00114ApplicationWB. Species reactivity: Human
Catalog No. 50-014-59132