Learn More
Cayman Chemical Necrostatin-1, Purity: ≥98%, CAS: 4311-88-0, 10 mg
SDP
Catalog No. 502953464
Encompass_Preferred
An inhibitor of RIP1 kinase that prevents the death of TNF-α-treated FADD-deficient Jurkat cells with an EC50 value of 490 nM
Catalog No. 50-295-3464 Supplier Cayman Chemical Supplier No. 1165810 MG
An inhibitor of RIP1 kinase that prevents the death of TNF-α-treated FADD-deficient Jurkat cells with an EC50 value of 490 nM