Learn More
Cayman Chemical ncrostatIn-2 10mg
SDP
Catalog No. 502953380
Encompass_Preferred
A potent inhibitor of necroptosis (EC50 = 50 nM); 4-fold more active than the (S)-enantiomer (EC50 = 230 nM)
Catalog No. 50-295-3380 Supplier Cayman Chemical Supplier No. 1165710 MG
A potent inhibitor of necroptosis (EC50 = 50 nM); 4-fold more active than the (S)-enantiomer (EC50 = 230 nM)