Learn More
Cayman Chemical ncrostatIn-34 10mg
SDP
Catalog No. 502916618
Encompass_Preferred
A RIPK1 inhibitor (IC50 = 5.5 µM); inhibits TNF-α-induced necroptosis in FADD-deficient Jurkat cells and L-929 cells (IC50s = 6.67 and 1.35 µM, respectively); inhibits AP20187- and Z-VA-DL-D(OMe)-FMK-induced necroptosis in L-929 cells expressing an FKBP and RIPK1 fusion protein at 10 µM
Catalog No. 50-291-6618 Supplier Cayman Chemical Supplier No. 3773510 MG
A RIPK1 inhibitor (IC50 = 5.5 µM); inhibits TNF-α-induced necroptosis in FADD-deficient Jurkat cells and L-929 cells (IC50s = 6.67 and 1.35 µM, respectively); inhibits AP20187- and Z-VA-DL-D(OMe)-FMK-induced necroptosis in L-929 cells expressing an FKBP and RIPK1 fusion protein at 10 µM