Learn More
Cayman Chemical ncrosulfonamIde 50mg
SDP
Catalog No. 502983601
Encompass_Preferred
Inhibits necroptosis by blocking MLKL, a key substrate of RIP3
Catalog No. 50-298-3601 Supplier Cayman Chemical Supplier No. 2084450 MG
Inhibits necroptosis by blocking MLKL, a key substrate of RIP3