Learn More
Cayman Chemical Necrosulfonamide-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1795144-22-7, 1 mg
SDP
Catalog No. 502910743
Encompass_Preferred
An internal standard for the quantification of necrosulfonamide by GC- or LC-MS
Catalog No. 50-291-0743 Supplier Cayman Chemical Supplier No. 347021 MG
An internal standard for the quantification of necrosulfonamide by GC- or LC-MS