Learn More
Cayman Chemical ncrosulfonamIde-d4 500ug
SDP
Supplier:  Cayman Chemical 34702500 UG
Encompass_Preferred
An internal standard for the quantification of necrosulfonamide by GC- or LC-MS
Catalog No. 50-291-2336