Learn More
Cayman Chemical Necrox-5 (methanesulfonate), Purity: ≥85%, CAS: 1383718-29-3, 10 mg
SDP
Catalog No. 502968438
Encompass_Preferred
A cell permeable inhibitor of necrosis that also demonstrates strong mitochondrial reactive oxygen species and ONOO- scavenging activity; localizes in mitochondria, selectively blocking oxidative stress-induced necrotic cell death (0.1 μM necrox-5 inhibited ~50% cell death in H9C2 cells exposed to 400 μM t-BHP)
Catalog No. 50-296-8438 Supplier Cayman Chemical Supplier No. 1727810 MG
A cell permeable inhibitor of necrosis that also demonstrates strong mitochondrial reactive oxygen species and ONOO- scavenging activity; localizes in mitochondria, selectively blocking oxidative stress-induced necrotic cell death (0.1 μM necrox-5 inhibited ~50% cell death in H9C2 cells exposed to 400 μM t-BHP)