Learn More
Medchemexpress LLC NCS-382 sodium | 131733-92-1 | MFCD11046018 | 99.0% | 240.23 | 1 MG
SDP
Catalog No. 5000452570
Encompass_Preferred
NCS-382 sodium is a potent GABA receptor antagonist, demonstrating anti-sedative and anti-hypnotic activities. It is a valuable tool for research related to neurological diseases and is intended for research use only.
  • Potent GABA receptor antagonist.
  • Exhibits anti-sedative and anti-hypnotic activities.
  • Suitable for research in neurological diseases.
Catalog No. 50-004-52570 Supplier Medchemexpress LLC Supplier No. HYW0086451MG
NCS-382 sodium is a potent GABA receptor antagonist, demonstrating anti-sedative and anti-hypnotic activities. It is a valuable tool for research related to neurological diseases and is intended for research use only.
  • Potent GABA receptor antagonist.
  • Exhibits anti-sedative and anti-hypnotic activities.
  • Suitable for research in neurological diseases.