Learn More
Cayman Chemical NCT-501 (hydrochloride), Purity: ≥98%, CAS: 2080306-22-3, 10 mg
SDP
Catalog No. 502967646
Encompass_Preferred
A potent, reversible, and selective inhibitor of ALDH1A1 (IC50 = 40 nM for the human form)
Catalog No. 50-296-7646 Supplier Cayman Chemical Supplier No. 1831910 MG
A potent, reversible, and selective inhibitor of ALDH1A1 (IC50 = 40 nM for the human form)