Learn More
Medchemexpress LLC NCT-502 100mg | 1542213-00-2 | 395.45 | C18H20F3N5S | 100 MG
SDP
Catalog No. 5000226976
Encompass_Preferred
NCT-502 is a small-molecule inhibitor of human phosphoglycerate dehydrogenase (PHGDH) supplied for research use. It is cytotoxic to PHGDH-dependent cancer cells and reduces glucose-derived serine production; reported IC50 against PHGDH is 3.7 μM. Molecular formula C18H20F3N5S, molecular weight 395.45 Da, reported purity 98.59%.
  • Potent PHGDH inhibitor (IC50 ≈ 3.7 μM).
  • Reduces glucose-derived serine production in cells.
  • Reported high purity suitable for biochemical assays.
  • Supplied as a 100 mg solid for laboratory research.
Catalog No. 50-002-26976 Supplier Medchemexpress LLC Supplier No. HY117240100MG
Add to Cart
Add to Cart
NCT-502 is a small-molecule inhibitor of human phosphoglycerate dehydrogenase (PHGDH) supplied for research use. It is cytotoxic to PHGDH-dependent cancer cells and reduces glucose-derived serine production; reported IC50 against PHGDH is 3.7 μM. Molecular formula C18H20F3N5S, molecular weight 395.45 Da, reported purity 98.59%.
  • Potent PHGDH inhibitor (IC50 ≈ 3.7 μM).
  • Reduces glucose-derived serine production in cells.
  • Reported high purity suitable for biochemical assays.
  • Supplied as a 100 mg solid for laboratory research.