Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NCT-502 10mg | 1542213-00-2 | 395.45 g·mol⁻1 | C18H20F3N5S | 10 MG
SDP

Catalog No. 5000227507
Encompass_Preferred

NCT-502 is a small-molecule inhibitor of human phosphoglycerate dehydrogenase (PHGDH). It reduces glucose-derived serine production and is cytotoxic to PHGDH-dependent cancer cells, making it suitable for biochemical and cellular studies of serine biosynthesis and metabolic vulnerabilities in cancer research.

  • PHGDH inhibitor (IC50 3.7 μM).
  • High purity (~98.59%).
  • Molecular weight 395.45 g·mol⁻1; formula C18H20F3N5S.
  • Soluble in DMSO (~130 mg/mL); sonication recommended for dissolution.
  • Stable when stored as powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Recommended frozen storage for solutions: -80°C (up to 2 years) or -20°C (up to 1 year).

Catalog No. 50-002-27507 Supplier Medchemexpress LLC Supplier No. HY11724010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NCT-502 is a small-molecule inhibitor of human phosphoglycerate dehydrogenase (PHGDH). It reduces glucose-derived serine production and is cytotoxic to PHGDH-dependent cancer cells, making it suitable for biochemical and cellular studies of serine biosynthesis and metabolic vulnerabilities in cancer research.

  • PHGDH inhibitor (IC50 3.7 μM).
  • High purity (~98.59%).
  • Molecular weight 395.45 g·mol⁻1; formula C18H20F3N5S.
  • Soluble in DMSO (~130 mg/mL); sonication recommended for dissolution.
  • Stable when stored as powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Recommended frozen storage for solutions: -80°C (up to 2 years) or -20°C (up to 1 year).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.