Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NCT-502 10mM 1mL | 1542213-00-2 | 1 ML
SDP

Catalog No. 5000228734
Encompass_Preferred

NCT-502 is a research-grade small molecule inhibitor of phosphoglycerate dehydrogenase (PHGDH). It is cytotoxic to PHGDH-dependent cancer cells and reduces glucose-derived serine production; reported IC50 against PHGDH is 3.7 μM. The compound is supplied as a light yellow to yellow solid and is offered as a 10 mM solution in DMSO (1 mL) and multiple powder sizes for laboratory research.

  • Mechanism: reversible PHGDH inhibitor with reported IC50 3.7 μM.
  • Purity: ~98.6%.
  • Appearance: light yellow to yellow solid.
  • Solubility: highly soluble in DMSO (≈130 mg/mL).
  • Storage: powder -20°C (long term) or 4°C; in solution -80°C or -20°C.
  • Pack formats: 1 mL DMSO solution (10 mM) and multiple mg quantities available.

Catalog No. 50-002-28734 Supplier Medchemexpress LLC Supplier No. HY11724010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

NCT-502 is a research-grade small molecule inhibitor of phosphoglycerate dehydrogenase (PHGDH). It is cytotoxic to PHGDH-dependent cancer cells and reduces glucose-derived serine production; reported IC50 against PHGDH is 3.7 μM. The compound is supplied as a light yellow to yellow solid and is offered as a 10 mM solution in DMSO (1 mL) and multiple powder sizes for laboratory research.

  • Mechanism: reversible PHGDH inhibitor with reported IC50 3.7 μM.
  • Purity: ~98.6%.
  • Appearance: light yellow to yellow solid.
  • Solubility: highly soluble in DMSO (≈130 mg/mL).
  • Storage: powder -20°C (long term) or 4°C; in solution -80°C or -20°C.
  • Pack formats: 1 mL DMSO solution (10 mM) and multiple mg quantities available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.