Learn More
TARGETMOL CHEMICALS INC NCT-502 25MG
Supplier: TARGETMOL CHEMICALS INC T1627725MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NCT-502 (N-(46-dimethylpyridin-2-yl)-4-[5-(trifluoromethyl)pyridin-2-yl]piperazine-1-carbothioamide) is a human phosphoglycerate dehydrogenase inhibitor cytotoxic to PHGDH-dependent cancer cells. It decreases glucose-derived serine production and it has an IC50 of 3.7 uM against PHGDH. purity: 99%