Learn More
Medchemexpress LLC HY-117240 5mg Medchemexpress, NCT-502 CAS:1542213-00-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1172405MG
Encompass_Preferred
Medchemexpress, HY-117240 5mg NCT-502 CAS:1542213-00-2 NCT-502 is a human phosphoglycerate dehydrogenase ( PHGDH ) inhibitor, cytotoxic to PHGDH-dependent cancer cells, and reduces glucose-derived serine production, with an IC 50 of 3.7 μM against PHGDH [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2517