Learn More
Selleck Chemical LLC NCT-503
SDP
Catalog No. 5000001871
Encompass_Preferred
NCT-503 is a phosphoglycerate dehydrogenase (PHGDH) inhibitor with the IC50 value of 2 5 M It is inactive against a panel of other dehydrogenases and shows minimal cross-reactivity in a panel of 168 GPCRs
Catalog No. 50-000-01871 Supplier Selleck Chemical LLC Supplier No. S861925MG
NCT-503 is a phosphoglycerate dehydrogenase (PHGDH) inhibitor with the IC50 value of 2 5 M It is inactive against a panel of other dehydrogenases and shows minimal cross-reactivity in a panel of 168 GPCRs