Learn More
Medchemexpress LLC NCT-503 | 1916571-90-8 | 99.7% | 408.48 g/mol | C20H23F3N4S | 200 MG
SDP
Catalog No. 5000247243
Encompass_Preferred
NCT-503 is a small-molecule inhibitor of phosphoglycerate dehydrogenase (PHGDH) with an IC50 of 2.5 μM, used as a research tool to probe serine biosynthesis pathways and PHGDH-dependent cancer biology in vitro and in vivo.
  • Potent PHGDH inhibition with IC50 2.5 μM.
  • High selectivity against other dehydrogenases and minimal GPCR cross-reactivity.
  • Demonstrated activity in PHGDH-dependent cell lines and tumor xenograft models.
  • Solid form with high purity suitable for biochemical and cellular assays.
  • Recommended storage conditions for long-term stability.
Catalog No. 50-002-47243 Supplier Medchemexpress LLC Supplier No. HY101966200MG
NCT-503 is a small-molecule inhibitor of phosphoglycerate dehydrogenase (PHGDH) with an IC50 of 2.5 μM, used as a research tool to probe serine biosynthesis pathways and PHGDH-dependent cancer biology in vitro and in vivo.
  • Potent PHGDH inhibition with IC50 2.5 μM.
  • High selectivity against other dehydrogenases and minimal GPCR cross-reactivity.
  • Demonstrated activity in PHGDH-dependent cell lines and tumor xenograft models.
  • Solid form with high purity suitable for biochemical and cellular assays.
  • Recommended storage conditions for long-term stability.