Learn More
TARGETMOL CHEMICALS INC NCT-504 5MG
SDP
Catalog No. 502580101
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NCT-504 a selective allosteric inhibitor of PIP4K(gamma) exhibits an IC50 value of 15.8 uM. It holds promise for the investigation of Huntington's disease. purity: 100%
Catalog No. 50-258-0101 Supplier TARGETMOL CHEMICALS INC Supplier No. T386055MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NCT-504 a selective allosteric inhibitor of PIP4K(gamma) exhibits an IC50 value of 15.8 uM. It holds promise for the investigation of Huntington's disease. purity: 100%