Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NCT-506 | 2231098-99-8 | 99.95% | 478.54 | 1 ML
SDP

Catalog No. 5000436618
Encompass_Preferred

NCT-506 is an orally bioavailable aldehyde dehydrogenase 1A1 (ALDH1A1) inhibitor with an IC50 of 7 nM. It is intended for research use only.

  • Inhibits ALDH1A1, hALDH1A3, and hALDH2 with IC50s of 0.007±0.001, 16.4±3.99, and 21.5 μM, respectively.
  • Decreases cell viability with an EC50 of 45.6 μM in OV-90 cells (3D culture) after 6 days.
  • Inhibits MIA PaCa-2, OV-90, and HT-29 cells in aldefluor cell-based assays.
  • Shows enhanced inhibitory effects when combined with Paclitaxel in SKOV-3-TR cells.

Catalog No. 50-004-36618 Supplier Medchemexpress LLC Supplier No. HY11227810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

NCT-506 is an orally bioavailable aldehyde dehydrogenase 1A1 (ALDH1A1) inhibitor with an IC50 of 7 nM. It is intended for research use only.

  • Inhibits ALDH1A1, hALDH1A3, and hALDH2 with IC50s of 0.007±0.001, 16.4±3.99, and 21.5 μM, respectively.
  • Decreases cell viability with an EC50 of 45.6 μM in OV-90 cells (3D culture) after 6 days.
  • Inhibits MIA PaCa-2, OV-90, and HT-29 cells in aldefluor cell-based assays.
  • Shows enhanced inhibitory effects when combined with Paclitaxel in SKOV-3-TR cells.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.