Learn More
ATCC NCTC clone 929 Areolar Fibroblast Mouse (Mus musculus(
Catalog No. 502382545
Encompass
NCTC clone 929 [L cell, L-929, derivative of Strain L] (connective mouse tissue) is the first clone of strain L, one of the first cell strains to be established in continuous culture. NCTC clone 929 can be used for cell line toxicity testing.
Catalog No. 50-238-2545 Supplier ATCC Supplier No. CCL1
 Restricted Item
Add to Cart
Add to Cart
NCTC clone 929 [L cell, L-929, derivative of Strain L] (connective mouse tissue) is the first clone of strain L, one of the first cell strains to be established in continuous culture. NCTC clone 929 can be used for cell line toxicity testing.