Learn More
TARGETMOL CHEMICALS INC ND-322 HCL 5MG
SDP
Catalog No. 502580545
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ND-322 HCl (ND 322 Hydrochloride) is a selective inhibitor of MT1-MMP and MMP2 and reduces in vitro melanoma cell growth migration and invasion. purity: 98%
Catalog No. 50-258-0545 Supplier TARGETMOL CHEMICALS INC Supplier No. T281455MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ND-322 HCl (ND 322 Hydrochloride) is a selective inhibitor of MT1-MMP and MMP2 and reduces in vitro melanoma cell growth migration and invasion. purity: 98%