Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ND-322 hydrochloride | 1333379-23-9 | 98.1% | 357.88 g/mol | C15H16ClNO3S2 | 1MG
SDP

Catalog No. 5000235166
Encompass_Preferred

ND-322 hydrochloride is the hydrochloride salt of ND-322, a potent, selective gelatinase (matrix metalloproteinase) inhibitor for research use. It is water-soluble and has been reported in preclinical studies to reduce melanoma tumor growth and delay metastatic dissemination. The compound is supplied as a light yellow solid and is intended for laboratory research applications.

  • potent and selective gelatinase (MMP) inhibitor
  • reported to reduce melanoma tumor growth and delay metastasis in preclinical models
  • hydrochloride salt, water-soluble; DMSO solubility reported as 100 mg/mL
  • purity 98.14%
  • molecular formula C15H16ClNO3S2; molecular weight 357.88 g/mol
  • appearance light yellow to yellow solid
  • storage sealed, away from moisture and light; in solvent -80°C (6 months), -20°C (1 month)

Catalog No. 50-002-35166 Supplier Medchemexpress LLC Supplier No. HY1249391MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ND-322 hydrochloride is the hydrochloride salt of ND-322, a potent, selective gelatinase (matrix metalloproteinase) inhibitor for research use. It is water-soluble and has been reported in preclinical studies to reduce melanoma tumor growth and delay metastatic dissemination. The compound is supplied as a light yellow solid and is intended for laboratory research applications.

  • potent and selective gelatinase (MMP) inhibitor
  • reported to reduce melanoma tumor growth and delay metastasis in preclinical models
  • hydrochloride salt, water-soluble; DMSO solubility reported as 100 mg/mL
  • purity 98.14%
  • molecular formula C15H16ClNO3S2; molecular weight 357.88 g/mol
  • appearance light yellow to yellow solid
  • storage sealed, away from moisture and light; in solvent -80°C (6 months), -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.