Learn More
Abcam ND-630, 5MG
Catalog No. 502205814
Encompass_Preferred
MW 569.6 Da, Purity >=98%. A potent, and reversible acetyl-CoA carboxylase (ACC) inhibitor; inhibits human ACC1 and ACC2 with IC₅₀ values of 2.1 and 6.1 nM, respectively. When administered chronically to Zucker diabetic fatty rats, ND-630 reduces hepatic steatosis, improves glucose-stimulated insulin secretion, and reduces hemoglobin A1c (0.9% reduction).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5814 Supplier Abcam Supplier No. AB2869675MG
MW 569.6 Da, Purity >=98%. A potent, and reversible acetyl-CoA carboxylase (ACC) inhibitor; inhibits human ACC1 and ACC2 with IC₅₀ values of 2.1 and 6.1 nM, respectively. When administered chronically to Zucker diabetic fatty rats, ND-630 reduces hepatic steatosis, improves glucose-stimulated insulin secretion, and reduces hemoglobin A1c (0.9% reduction).
The product is subject to the following: Abcam Restricted Use Statement