Learn More
Aviva Systems Biology Corp ND5 Antibody - N-terminal region (ARP50254_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50254_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ND5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ND5. Alternate names: MTND5ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53623