Learn More
Aviva Systems Biology Corp ND6 Antibody (ARP50255_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50255_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ND6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ND6. Alternate names: MTND6ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-53624