Learn More
Selleck Chemical LLC ND646-5mg
SDP
Supplier:  Selleck Chemical LLC S83775MG
Encompass_Preferred
ND-646 is an allosteric inhibitor of the ACC (Acetyl-coA carboxylase) enzymes that prevents ACC subunit dimerization to suppress fatty acid synthesis with IC50 of 3.5 nM and 4.1 nM for hACC1 and hACC2, respectively.
Catalog No. 50-238-9702