Learn More
Trajan Scientific ND9 SCREW CAP 2CV MAG GOLD
Catalog No. NC1526476
Encompass
ND9 Screw Cap 2CV Mag GoldMagnetic, ND9 Cap for 2mL ScrewTop VialsGold, 1.0mm Silicone/PTFE/Starburst-Slitted septa
Catalog No. NC1526476 Supplier Trajan Scientific Supplier No. CP.09SP10SSTC CAP
ND9 Screw Cap 2CV Mag GoldMagnetic, ND9 Cap for 2mL ScrewTop VialsGold, 1.0mm Silicone/PTFE/Starburst-Slitted septa