Learn More
Cayman Chemical nDD8 InhIbItr M22 5mg
SDP
Catalog No. 502916539
Encompass_Preferred
An inhibitor of NEDD8-activating enzyme; inhibits NEDDylation of UBC12 in a cell-free assay using recombinant human NAE; selectively inhibits NEDDylation over SUMOylation and ubiquitination in A549 cells; inhibits the growth of A549 cells (GI50 = 5.5 µM); inhibits tumor growth in an AGS gastric adenocarcinoma mouse xenograft model at 60 mg/kg
Catalog No. 50-291-6539 Supplier Cayman Chemical Supplier No. 377565 MG
Add to Cart
Add to Cart
An inhibitor of NEDD8-activating enzyme; inhibits NEDDylation of UBC12 in a cell-free assay using recombinant human NAE; selectively inhibits NEDDylation over SUMOylation and ubiquitination in A549 cells; inhibits the growth of A549 cells (GI50 = 5.5 µM); inhibits tumor growth in an AGS gastric adenocarcinoma mouse xenograft model at 60 mg/kg