Learn More
Cayman Chemical N-decanoyl-L-Homoserine lactone, Purity: ≥96%, CAS: 177315-87-6, 25 mg
SDP
Catalog No. 502889320
Encompass_Preferred
A small diffusible signaling molecule involved in quorum sensing, controlling gene expression, and affecting cellular metabolism; applications for this molecule include regulation of virulence and exoproteases
Catalog No. 50-288-9320 Supplier Cayman Chemical Supplier No. 1001120125 MG
A small diffusible signaling molecule involved in quorum sensing, controlling gene expression, and affecting cellular metabolism; applications for this molecule include regulation of virulence and exoproteases